missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112690
This item is not returnable.
View return policy
Description
RGL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Probable guanine nucleotide exchange factor. Putative effector of Ras and/or Rap. Associates with the GTP-bound form of Rap 1A and H-Ras in vitro.
Specifications
| RGL2 | |
| Polyclonal | |
| Unconjugated | |
| RGL2 | |
| GDS-related protein; HKE1.5; KE1.5; RAB2L; ral guanine nucleotide dissociation stimulator like 2; ral guanine nucleotide dissociation stimulator,-like 2; ral guanine nucleotide dissociation stimulator-like 2; RalGDS-like 2; ralGDS-like factor; Ras-associated protein RAB2L; Rgl2; Rgt2; Rlf | |
| Rabbit | |
| Protein G | |
| RUO | |
| 19732, 294283, 5863 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O15211, Q61193 | |
| RGL2 | |
| Recombinant Human Ral guanine nucleotide dissociation stimulator-like 2 protein (145-328AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction