missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68676
This item is not returnable.
View return policy
Description
RGL2 Polyclonal antibody specifically detects RGL2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| RGL2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FDKRRKEFAVLSELRRLQNECRGYNLQPDHDIQRWLQGLRPLTEAQSHRVSCEVEPPGSSDPPAPRVLRPTLVISQWA | |
| 100 μg | |
| Signal Transduction | |
| 5863 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction