missing translation for 'onlineSavingsMsg'
Learn More

RGL2 Antibody (4D10), Novus Biologicals™

Product Code. 18378868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378868 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378868 Supplier Novus Biologicals Supplier No. H00005863M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RGL2 Monoclonal antibody specifically detects RGL2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGL2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4D10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32681
Gene Alias GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor
Host Species Mouse
Immunogen RGL2 (AAH32681, 644 a.a. ~ 743 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PGASDCRIIRVQMELGEDGSVYKSILVTSQDKAPSVISRVLKKNNRDSAVASEYELVQLLPGERELTIPASANVFYAMDGASHDFLLRQRRRSSTATPGV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5863
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.