missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104054
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RGL1.
Ral GDS (Ral guanine nucleotide dissociation stimulator) is a guanine nucleotide exchange factor (GEF) that activates Ral and is implicated in oncogenic Ras-induced cell transformation. RGL1 (ral guanine nucleotide dissociation stimulator-like 1), also known as RGL or RalGDS-like 1, is a 768 amino acid protein that is a putative GEF. Strongly expressed in brain, heart, spleen, kidney and testis, RGL1 is a downstream effector protein that is involved in Ras and Ral signaling pathways. RGL1 contains an N-terminal Ras-GEF domain and a C-terminal Ras-interacting domain that interacts with the GTP-bound form of Ras through its effector loop. Due to its similarity to Ral GDS, RGL1 may be implicated in carcinogenesis. Two isoforms exist due to alternative splicing events.
Specifications
| RGL1 | |
| Polyclonal | |
| Unconjugated | |
| RGL1 | |
| KIAA0959; mKIAA0959; ral guanine nucleotide dissociation stimulator like 1; ral guanine nucleotide dissociation stimulator,-like 1; ral guanine nucleotide dissociation stimulator-like 1; RalGDS-like 1; RGL; Rgl1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19731, 23179, 289080 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q60695, Q9NZL6 | |
| RGL1 | |
| A synthesized peptide derived from human RGL1(Accession Q9NZL6), corresponding to amino acid residues P532-K582. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction