missing translation for 'onlineSavingsMsg'
Learn More

CABCOCO1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15828543
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15828543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15828543 Supplier Invitrogen Supplier No. PA570132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CABCOCO1 Polyclonal Antibody

CABCOCO1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CABCOCO1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CABCOCO1
Gene Accession No. 0
Gene Alias bA63A2.1; C10orf107; CABCOCO1; chromosome 10 open reading frame 107; Ciliary-associated calcium-binding coiled-coil protein 1; RGD1306739; similar to RIKEN cDNA 1700040L02; uncharacterized protein C10orf107; uncharacterized protein C10orf107 homolog
Gene Symbols CABCOCO1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat RGD1306739.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361834
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.