missing translation for 'onlineSavingsMsg'
Learn More

RGC32 Antibody (3B9), Novus Biologicals™

Product Code. 18378779 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378779 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378779 Supplier Novus Biologicals Supplier No. H00028984M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RGC32 Monoclonal antibody specifically detects RGC32 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGC32
Applications ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 3B9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_054778
Gene Alias bA157L14.2, chromosome 13 open reading frame 15, KIAA0564, RGC-32MGC87338, RGC32response gene to complement 32 protein
Host Species Mouse
Immunogen C13orf15 (NP_054778.1, 1 a.a. ~ 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASM
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 28984
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.