missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGB-286638, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
RGB-286638 is a CDK inhibitor that inhibits the kinase activity of cyclin T1-CDK9, cyclin B1-CDK1, cyclin E-CDK2, cyclin D1-CDK4, cyclin E-CDK3, and p35-CDK5 with IC50s of 1, 2, 3, 4, 5 and 5 nM, respectively
Specifications
Specifications
| Chemical Name or Material | RGB-286638 |
| Physical Form | Solid |
| CAS | 784210-87-3 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C29H37Cl2N7O4 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : ≥ 150 mg/mL (242.50 mM) |
| SMILES | O=C(NN1CCOCC1)NC2=CC=CC(C3=C4C(C5=CC=C(CN6CCN(CCOC)CC6)C=C5)=NN3)=C2C4=O.[H]Cl.[H]Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction