missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGAG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551985
This item is not returnable.
View return policy
Description
Immunogen sequence: ISVTQEGSPL HANFPRFLDE IRKEFCGPIP PRVAKKAIRK LKQGHCTLGS YADAFQFLAQ FLSWDDCRLQ NQFLKGLSEF FRKELLWSTE MADLDELILE CVE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 27%.
Specifications
| RGAG4 | |
| Polyclonal | |
| Unconjugated | |
| RTL5 | |
| 5-Mar; 6430402L03Rik; gag-like protein; Kiaa2001; LOC100911832; MAR5; Mart5; mKIAA2001; retrotransposon gag domain containing 4; retrotransposon gag domain-containing protein 4; retrotransposon gag domain-containing protein 4-like; Retrotransposon Gag-like protein 5; RGAG4; Rtl5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100911832, 331474, 340526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5DTT4, Q5HYW3 | |
| RTL5 | |
| Recombinant protein corresponding to Human RGAG4. Recombinant protein control fragment (Product #RP-92257). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu