missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGAG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £435.00
Specifications
| Antigen | RGAG1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18418630
|
Novus Biologicals
NBP1-82914-25ul |
25 μL |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18285436
|
Novus Biologicals
NBP1-82914 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGAG1 Polyclonal specifically detects RGAG1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RGAG1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 57529 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TSTLLMRDTASGVMSCPQMRSLASGALSKPLMTPKASGTMFTEKMTTTASEAMPTLLMRDTVSGALSMPQMTDTASGGLSASLMRDTASGAMSTSQMTATVSGGMSMPLMRAQDPGVMPASLMRAKVSGKMLSQPMSTQDPGGMSM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| KIAA1318MGC142230, Mar9, Mart9, retrotransposon gag domain containing 1, retrotransposon gag domain-containing protein 1, tumor antigen BJHCC23, Tumor antigen BJ-HCC-23 | |
| RGAG1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title