missing translation for 'onlineSavingsMsg'
Learn More

RG7834, MedChemExpress

Product Code. 16467637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16467637

Brand: Medchem Express HY117650A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG7834 (RO 7020322) is a highly selective and orally bioavailable HBV inhibitor, potently inhibits HBV antigens (both HBsAg and HBeAg) and HBV DNA, with IC50s of 2.8, 2.6, and 3.2 nM, respectively, in dHepaRG Cells.

RG7834 (RO 7020322) is a highly selective and orally bioavailable HBV inhibitor, potently inhibits HBV antigens (both HBsAg and HBeAg) and HBV DNA, with IC50s of 2.8, 2.6, and 3.2 nM, respectively, in dHepaRG Cells.

Specifications

Chemical Name or Material RG7834
CAS 2072057-17-9
Color White
Purity Grade Notes Research
Molecular Formula C22H27NO6
Quantity 5 mg
Synonym RO 7020322
Solubility Information DMSO : 125 mg/mL (311.37 mM; Need ultrasonic)
SMILES O=C(O)C1=CN2[C@@H](CC3=CC(OCCCOC)=C(C=C3C2=CC1=O)OC)C(C)C
Molecular Weight (g/mol) 401.45
Formula Weight 401.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.