missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RG7800 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
RG7800 hydrochloride is an orally active SMN2 splicing modulator, with EC1.5xs of 23 nM and 87 nM for SMN2 splicing and SMN protein
Specifications
Specifications
| Chemical Name or Material | RG7800 hydrochloride |
| Physical Form | Solid |
| Color | Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H28N6O.xHCl |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | RO6885247 hydrochloride |
| Solubility Information | H2O : ≥ 113.5 mg/mL |
| SMILES | CC1=CC(C2CCN(C)CC2)=CN3C1=NC(C4=NN(C=C(C)N=C5CC)C5=C4)=CC3=O.[xHCl] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction