missing translation for 'onlineSavingsMsg'
Learn More

RG7713, MedChemExpress

Product Code. 16405468
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405468

Brand: Medchem Express HY12981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a).

RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a).

Specifications

Chemical Name or Material RG7713
CAS 920022-47-5
Color White
Purity Grade Notes Research
Molecular Formula C25H28ClN3O2
Quantity 10 mg
Synonym RO5028442
Solubility Information DMSO : 20 mg/mL (45.67 mM; Need ultrasonic)
SMILES O=C(C1=CN(CCN(C)C)C2=C1C=CC(Cl)=C2)N3CCC4(C(C=CC=C5)=C5CO4)CC3
Molecular Weight (g/mol) 437.96
Formula Weight 437.96
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.