missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RG108

Product Code. 17140042
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140042 5 mg 5mg
17150042 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140042 Supplier STEMCELL Technologies Supplier No. 72212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epigenetic modifier; Inhibits DNA methyltransferase (DNMT)

  • RG108 is an epigenetic modifier that inhibits DNA methyltransferase (IC50 = 115 nM)
  • RG108 is a non-nucleoside inhibitor that acts by direct binding to the methyltransferase enzyme whereby it blocks the enzyme active site
  • (Brueckner et al.), Stresemann et al.)
  • Enhances reprogramming efficiency of human and mouse somatic cells to induced pluripotent stem (iPS) cells (Mali et al.), Pasha et al.), Shi et al.)

Specifications

CAS 48208-26-0
Synonym N-Phthalyl-L-Tryptophan
Quantity 5 mg
Molecular Formula C19H14N2O4
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.