missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RG1 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Product Code. 30284629
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30284629 100 μg 100µg
30284633 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284629 Supplier Invitrogen™ Supplier No. MA558949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term 19G9.

RG1 is a gene that has primarily been studied in the context of Ginsenoside Rg1, a bioactive compound derived from the roots of Panax ginseng. Ginsenoside Rg1 is known for its neuroprotective effects and regulatory role in immune responses. It modulates various signaling pathways, including the phosphatidylinositol 3-kinase (PI3K)/Akt pathway, which is crucial for cell survival and growth. Additionally, Rg1 has been shown to inhibit apoptotic pathways mediated by endoplasmic reticulum stress and the c-Jun N-terminal kinase (JNK) pathway in neurodegenerative disease models, potentially offering therapeutic benefits in conditions like Alzheimer's disease. Furthermore, Rg1 influences the activation of astrocytes through long non-coding RNAs (lncRNAs) and microRNAs, contributing to the repair of spinal cord injuries. Overall, RG1, through its association with Ginsenoside Rg1, plays a significant role in cellular protection and modulation across various biological systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RG1 Biosimilar
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.