missing translation for 'onlineSavingsMsg'
Learn More

RG-12525, MedChemExpress

Product Code. 16496217
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16496217

Brand: Medchem Express HY101676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG-12525 is a a specific, competitive and orally effective antagonist of the peptidoleukotrienes, LTC4, LTD4 and LTE4, inhibiting LTC4-, LTD4- and LTE4-inducd guinea pig parenchymal strips contractions, with IC50s of 2.6 nM, 2.5 nM and 7 nM, respectively; RG-12525 is also a peroxisome proliferator-activated receptor gamma (PPAR-gamma) agonist with IC50 of appr 60 nM and a potent inhibitor of CYP3A4, with a Ki value of 0.5 μM.

RG-12525 is a a specific, competitive and orally effective antagonist of the peptidoleukotrienes, LTC4, LTD4 and LTE4, inhibiting LTC4-, LTD4- and LTE4-inducd guinea pig parenchymal strips contractions, with IC50s of 2.6 nM, 2.5 nM and 7 nM, respectively

Specifications

Chemical Name or Material RG-12525
CAS 120128-20-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H21N5O2
Quantity 5 mg
Synonym NID 525
Solubility Information DMSO : 25 mg/mL (59.04 mM; Need ultrasonic)
SMILES C1(COC2=CC=C(OCC3=CC=CC=C3CC4=NN=NN4)C=C2)=NC5=CC=CC=C5C=C1
Molecular Weight (g/mol) 423.47
Formula Weight 423.47
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.