missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFXAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 491034
This item is not returnable.
View return policy
Description
RFXAP Polyclonal Antibody for ChIP
Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated ankyrin-containing protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group D. Transcript variants utilizing different polyA signals have been found for this gene.
Specifications
| RFXAP | |
| Polyclonal | |
| Unconjugated | |
| RFXAP | |
| 5730495K23Rik; regulatory factor X associated protein; regulatory factor X-associated protein; RFX DNA-binding complex 36 kDa subunit; RFXAP; RFX-AP; RFX-associated protein; si:ch211-170h2.4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5994 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay | |
| 1 mg/mL | |
| PBS with 0.05% ProClin 300, 0.05% sodium azide | |
| O00287 | |
| RFXAP | |
| Raised against the recombinant RFXAP protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction