missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFXAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58217
This item is not returnable.
View return policy
Description
RFXAP Polyclonal specifically detects RFXAP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| RFXAP | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| regulatory factor X-associated protein, RFX DNA-binding complex 36 kDa subunit, RFX-associated protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| RFXAP | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NVKLEESADNILSIVKQRTGSFGDRPARPTLLEQVLNQKRLSLLRSPEVVQFLQKQQQLLNQQVLEQRQQQFPGT | |
| 100 μL | |
| DNA Repair, Stem Cell Markers, Transcription Factors and Regulators | |
| 5994 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction