missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528878
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RFX6 is a member of the regulatory factor X.
Specifications
| RFX6 | |
| Polyclonal | |
| Unconjugated | |
| RFX6 | |
| 4930572O07Rik; dJ955L16.1; DNA-binding protein RFX6; MTCHRS; MTFS; Regulatory factor X 6; regulatory factor X domain containing 1; regulatory factor X domain-containing protein 1; regulatory factor X, 6; regulatory factor X6; RFX6; RFXDC1; si:dkeyp-93d12.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222546 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.99 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q8HWS3 | |
| RFX6 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human RFX6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction