missing translation for 'onlineSavingsMsg'
Learn More

RFX6 Antibody [Biotin], Novus Biologicals™

Product Code. 30733613 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733613 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733613 Supplier Novus Biologicals Supplier No. AF7780B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

RFX6 Polyclonal antibody specifically detects RFX6 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX6
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias dJ955L16.1, DNA-binding protein RFX6, MGC33442, Regulatory factor X 6, Regulatory factor X domain-containing protein 1, regulatory factor X, 6, RFXDC1regulatory factor X domain containing 1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human RFX6, Lys324-Thr511, Accession # Q8HWS3
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222546
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.