missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12137-1-AP-150UL
This item is not returnable.
View return policy
Description
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. Directly associates with HAX-1, through binding to its C-terminal region. May also be involved in the regulation of gene expression.Specifications
| RFX5 | |
| Polyclonal | |
| Unconjugated | |
| RFX5 | |
| DNA binding protein RFX5, Regulatory factor X 5, RFX5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5993 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P48382 | |
| RFX5 | |
| RFX5 Fusion Protein Ag2783 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction