Learn More
RFX5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NB100-195
Product Code. 18717223
  • £452.00 / 0.10mL
Description
RFX5 Polyclonal specifically detects RFX5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.0 mg/mL
DNA-binding protein RFX5, Regulatory factor X 5, regulatory factor X, 5 (influences HLA class II expression)
Affinity Purified
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
RFX5