missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: QYMAMRQQPM QQKQRYKPMQ KVDGVADGFT GSGQQTGTSV EQTVIAQSQH HQQFLDASRA LPEFGEVEIS SLPDGTTFED IKSLQSLYRE HCEAILDVVV NLQFSLIEKL WQTFWRYSPS TPTDGTTITE SSNLSE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with other RFX family members. Two transcript variants encoding different isoforms have been described for this gene, and at least one of the variants utilizes alternative polyadenylation signals.
Specifications
Specifications
| Antigen | RFX3 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RFX3 |
| Gene Accession No. | P48380 |
| Gene Alias | bA32F11.1; C230093O12Rik; DNA binding protein RFX3; MRFX3; Regulatory factor X 3; regulatory factor X, 3 (influences HLA class II expression); regulatory factor X3; Rfx2; RFX3; Transcription factor RFX3 |
| Gene Symbols | RFX3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
