missing translation for 'onlineSavingsMsg'
Learn More

RFX3 Antibody (OTI3F7), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18418186 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418186 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418186 Supplier Novus Biologicals Supplier No. NBP273865AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RFX3 Monoclonal antibody specifically detects RFX3 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX3
Applications Western Blot
Classification Monoclonal
Clone OTI3F7
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias bA32F11.1, DNA binding protein RFX3, MGC87155, Regulatory factor X 3, regulatory factor X, 3 (influences HLA class II expression), transcription factor RFX3
Host Species Mouse
Immunogen Full length recombinant protein of human RFX3 (NP_602304) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5991
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.