missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550023
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total RFX2 protein.
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with other RFX family members. This protein can bind to cis elements in the promoter of the IL-5 receptor alpha gene. Two transcript variants encoding different isoforms have been described for this gene, and both variants utilize alternative polyadenylation sites. [provided by RefSeq, Jul 2008].
Specifications
| RFX2 | |
| Polyclonal | |
| Unconjugated | |
| RFX2 | |
| DNA binding protein RFX2; DNA-binding protein RFX2; FLJ14226; HLA class II regulatory factor RFX2; regulatory factor X 2; regulatory factor X, 2 (influences HLA class II expression); regulatory factor X2; RFX2; trans-acting regulatory factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5990 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48378 | |
| RFX2 | |
| Synthesized peptide derived from internal of human RFX2 (aa 661-710). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction