missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-82031G
This item is not returnable.
View return policy
Description
RFX1 Polyclonal antibody specifically detects RFX1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| RFX1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 488 | |
| EFC, EF-C, Enhancer factor C, MHC class II regulatory factor RFX, MHC class II regulatory factor RFX1, Regulatory factor X 1, regulatory factor X, 1 (influences HLA class II expression), RFX, trans-acting regulatory factor 1, Transcription factor RFX1 | |
| RFX1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RFX1. The immunogen is located within the last 50 amino acids of RFX1. Amino Acid Squence: AGGESPALGPETLEPPAK | |
| 0.1 mL | |
| Immunology | |
| 5989 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction