missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFWD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | RFWD3 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18234145
|
Novus Biologicals
NBP2-58819 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652807
|
Novus Biologicals
NBP2-58819-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RFWD3 Polyclonal specifically detects RFWD3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RFWD3 | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| 55159 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NHTIPASSLHSMTNFISGLQRLHGMLEFLRPSSSNHSVGPMRTRRRVSASRRARAGGSQRTDSARLRAPLDAYFQVSRTQPDLPATTYDSETRNPVSE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| EC 6.3.2.-, FLJ10520, ring finger and WD repeat domain 3, RING finger and WD repeat domain-containing protein 3, RING finger protein 201, RNF201E3 ubiquitin-protein ligase RFWD3 | |
| RFWD3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title