missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFTN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17854-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| RFTN2 | |
| Polyclonal | |
| Unconjugated | |
| RFTN2 | |
| C2orf11, Raft linking protein 2, Raftlin 2, raftlin family member 2, RFTN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 130132, 363231, 74013 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q52LD8, Q8CHX7 | |
| Rftn2 | |
| RFTN2 Fusion Protein Ag12307 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction