missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFPL4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104098
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RFPL4B.
RFPL4B gene ontology (GO) annotation includes metal ion binding.
Specifications
| RFPL4B | |
| Polyclonal | |
| Unconjugated | |
| RFPL4B | |
| ret finger protein like 4B; ret finger protein-like 4B; RFPL4B; RING finger protein 211; RNF211 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 442247 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ZWI9 | |
| RFPL4B | |
| A synthesized peptide derived from human RFPL4B(Accession Q6ZWI9), corresponding to amino acid residues F93-S143. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction