missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFPL4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561227
This item is not returnable.
View return policy
Description
Immunogen sequence: PDGEGLLCRF CSVVSQKDDI KPKYKLRALV SIIK Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 53%.
RFPL4A gene ontology (GO) annotation includes metal ion binding and nucleus.
Specifications
| RFPL4A | |
| Polyclonal | |
| Unconjugated | |
| RFPL4A | |
| ret finger protein like 4A; ret finger protein-like 4; ret finger protein-like 4A; RFPL4; Rfpl4a; RING finger protein 210; RNF210 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 342931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NLU0 | |
| RFPL4A | |
| Recombinant protein corresponding to Human RFPL4A. Recombinant protein control fragment (Product #RP-98934). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction