missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFPL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567240
This item is not returnable.
View return policy
Description
Immunogen sequence: QEASSCPVCSD YLEKPMSLEC GCTVC Highest antigen sequence identity to the following orthologs - mouse 54%, rat 50%.
RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
Specifications
| RFPL3 | |
| Polyclonal | |
| Unconjugated | |
| RFPL3 | |
| ret finger protein like 3; ret finger protein-like 3; RFPL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75679 | |
| RFPL3 | |
| Recombinant Human RFPL3. Recombinant protein control fragment (Product #RP-105661). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction