Learn More
Invitrogen™ RFPL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588495
Description
Immunogen sequence: MAALFQEASS CPVCSDYLEK PMSLECGCTV CLKCINSLQK EPHGEDLLCC CCSMVSQRNK IRPNRQLERL VSHIKELEPK LKKILQMNPR MRKFQVDMTL DADTANNFLL ISDDLRSVRS GLITQNRQDL AERFDVSVCI LGSPRFTCGR HYWEVDVGTS TEWDLGVCRE SVHCKGKIQL TTELGFWTVS LRDGSRLSAS TVPLTFLLVD RKLQRVGIFL DMGMQNVSFF DAESGSHVYT FRSVSAEEPL RPFLAPSIPP NGDQGVLSIC PLMNSGTTDA PVRPGEAK; Positive Samples: U-87MG, HeLa.
RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
Specifications
| RFPL3 | |
| Polyclonal | |
| Unconjugated | |
| RFPL3 | |
| ret finger protein like 3; ret finger protein-like 3; RFPL3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10738 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75679 | |
| RFPL3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-288 of human RFPL3 (NP_0065951). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.