missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFPL3 Monoclonal Antibody (OTI2G4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525966
This item is not returnable.
View return policy
Description
RFPL3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
Specifications
| RFPL3 | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75679 | |
| RFPL3 | |
| Full length human recombinant protein of RFPL3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2G4 | |
| Unconjugated | |
| RFPL3 | |
| ret finger protein like 3; ret finger protein-like 3; RFPL3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10738 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction