missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL2 Polyclonal Antibody

RFPL2 Polyclonal Antibody
Brand: Invitrogen PA598117
This item is not returnable.
View return policy
Description
RFPL2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| RFPL2 | |
| Polyclonal | |
| Unconjugated | |
| RFPL2 | |
| ret finger protein like 2; ret finger protein-like 2; RFPL2; RING finger protein 79; RNF79 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10739 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75678 | |
| RFPL2 | |
| Recombinant Human Ret finger protein-like 2 protein (99-378AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction