missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540832
This item is not returnable.
View return policy
Description
Peptide sequence: DTLETSKRKS LQLLTKDSDK VKEFFEKLQH TLDQKKNEIL SDFETMKLAV Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
The protein encoded by the RFP2 gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies near the nucleus, but its function is unknown. The gene is located on chromosome 13 within the minimal deletion region for B-cell chronic lymphocytic leukemia.
Specifications
| RFP2 | |
| Polyclonal | |
| Unconjugated | |
| TRIM13 | |
| 3110001L12Rik; B-cell chronic lymphocytic leukemia tumor suppressor Leu5; CAR; CLL-associated RING finger; DLEU5; E3 ubiquitin-protein ligase TRIM13; LEU5; Leukemia-associated protein 5; putative tumor suppressor RFP2; Ret finger protein 2; Rfp2; RING finger protein 77; RING-type E3 ubiquitin transferase TRIM13; RNF77; TRIM13; tripartite motif containing 13; tripartite motif protein 13; tripartite motif-containing 13; tripartite motif-containing protein 13; tripartite motif-containing protein 13; E3 ubiquitin-protein ligase TRIM13; Unknown (protein for MGC:134296) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10206 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60858 | |
| TRIM13 | |
| Synthetic peptide directed towards the middle region of human RFP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction