missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFP Monoclonal Antibody (RF5R)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA515257

Product Code. 11325243

  • £244.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-15257 recognizes native and denatured forms of RFP (Red Fluorescent Protein) and its many variants: DsRed, mCherry, mOrange, etc. MA5-15257 has successfully been used in immunoprecipitation, ELISA, Western blotting, immunohistochemistry, immunocytochemistry and immunofluorescence applications. MA5-15257 may be stored at 4°C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20°C, avoiding freeze/thaw cycles.'.

Red Fluorescent Protein (RFP) is isolated from sea anemone (Discosoma sp, Uniprot ID: Q9U6Y8), and commonly used as a molecular tag to study protein localization, reporter gene expression, and protein-protein interactions. Although it exists as a homotetramer, monomeric variants have been constructed for use in various applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RFP
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Mouse
Protein A
RUO
Tag
Antibody
IgG1
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
RF5R
Unconjugated
DsRed2; mRFP; red fluorescent; Red fluorescent protein; RFP; RFP tag
RFP from the Discosoma sea anemone N-terminal peptide-KLH conjugated.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.