missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFP Recombinant Rabbit Monoclonal Antibody (HL2028)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547262
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Red Fluorescent Protein (RFP) is isolated from sea anemone (Discosoma sp, Uniprot ID: Q9U6Y8), and commonly used as a molecular tag to study protein localization, reporter gene expression, and protein-protein interactions. Although it exists as a homotetramer, monomeric variants have been constructed for use in various applications.
Specifications
| RFP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Tag | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2028 | |
| Unconjugated | |
| DsRed2; mRFP; red fluorescent; Red fluorescent protein; RFP; RFP tag | |
| Full length RFP recombinant protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction