missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45840-0.05MG
This item is not returnable.
View return policy
Description
RFP Polyclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot.
Specifications
| RFP | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 30% glycerol, 0.5% BSA with 0.01% Thimerosal | |
| Rabbit | |
| 0.05 mg | |
| Non-species specific | |
| IgG |
| Western Blot | |
| 0.50 mg/mL | |
| Western Blot | |
| dsRedGene Symbol, Recombinant RFP Protein, Red Fluorescent Protein | |
| Recombinant RFP expressed from E. coli. | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction