missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFP Antibody (1610), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43713
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
RFP Monoclonal specifically detects RFP in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| RFP | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| Mouse | |
| 27 kDa | |
| 0.1 mL | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1610 | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:500-1:1000 | |
| DSRED, GFP-like chromoprotein, Red Fluorescent Protein, RFP | |
| Recombinant protein conjugated synthetic peptide encompassing a sequence within the center region of RFP. The exact sequence is proprietary. | |
| Protein G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction