missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFNG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565105
This item is not returnable.
View return policy
Description
Immunogen sequence: SLRPDDVFIAV KTTRKNHGPR LRLLLRTWIS RARQQTFIFT DGDDPELELQ GGDRVINTNC SAVRTRQALC CKMSVEYDKF IESGRKWFCH VDDDNYVNAR SLLHLLSSFS PSQDVYLGRP SLDHPIEATE RV Highest antigen sequence identity to the following orthologs - mouse 87%, rat 88%.
Glycosyltransferase that initiates the elongation of O- linked fucose residues attached to EGF-like repeats in the extracellular domain of Notch molecules. May be involved in limb formation and in neurogenesis.
Specifications
| RFNG | |
| Polyclonal | |
| Unconjugated | |
| RFNG | |
| Beta-1,3-N-acetylglucosaminyltransferase radical fringe; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; radical fringe gene homolog; radical fringe homolog; Rfng; RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5986 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y644 | |
| RFNG | |
| Recombinant Human RFNG. Recombinant protein control fragment (Product #RP-105861). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction