missing translation for 'onlineSavingsMsg'
Learn More

RFNG Antibody (6C7), Novus Biologicals™

Product Code. 18335729 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335729 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335729 Supplier Novus Biologicals Supplier No. H00005986M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RFNG Monoclonal antibody specifically detects RFNG in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFNG
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6C7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Alias beta-1,3-N-acetylglucosaminyltransferase radical fringe, EC 2.4.1.222, O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, radical fringe (Drosophila) homolog, radical fringe homolog, radical fringe homolog (Drosophila), RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Host Species Mouse
Immunogen RFNG (AAC51359.1, 82 a.a. ~ 190 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LGSFMSTAEQVRLPDDCTVGYIVEGLLGARLLHSPLFHSHLENLQRLPPDTLLQQVTLSHGGPENPQNVVNVAGGFSLHQDPTRFKSIHCLLYPDTDWCPRQKQGAPTS
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5986
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.