missing translation for 'onlineSavingsMsg'
Learn More

RFK Antibody, DyLight 550, Novus Biologicals™

Product Code. 18362666 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362666 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362666 Supplier Novus Biologicals Supplier No. NBP300208R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RFK Polyclonal specifically detects RFK in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFK
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias ATP:riboflavin 5'-phosphotransferase, EC 2.7.1.26, Flavokinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RFK (Q969G6; Met1-His155).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55312
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.