missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFFL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12687-1-AP-20UL
This item is not returnable.
View return policy
Description
Has E3 ubiquitin protein ligase activity. Regulates the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. Has anti-apoptotic activity. May bind phosphatidylinositol phosphates.Specifications
| RFFL | |
| Polyclonal | |
| Unconjugated | |
| RFFL | |
| CARP 2, Caspase regulator CARP2, Fring, RFFL, RIFIFYLIN, RING finger protein 189, RING finger protein 34 like, RNF189, RNF34L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117584, 282844, 67338 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZQM0, Q8CIN9, Q8WZ73 | |
| Rffl | |
| RFFL Fusion Protein Ag3380 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction