missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFESD Antibody (OTI2A7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73860B
This item is not returnable.
View return policy
Description
RFESD Monoclonal specifically detects RFESD in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RFESD | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| RFESD | |
| Full length recombinant protein of human RFESD (NP_775498) produced in HEK293T cells. | |
| 0.1 mL | |
| 317671.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2A7 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Rieske (Fe-S) domain containing, Rieske domain-containing protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction