Learn More
RFC5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28671-1-AP-150UL
This item is not returnable.
View return policy
Description
The RFC5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RFC5. This antibody recognizes human antigen. The RFC5 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| RFC5 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| 150 μL | |
| 5985 | |
| IgG |