missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103494
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RFC5.
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 36 kD subunit. This subunit can interact with the C-terminal region of PCNA. It forms a core complex with the 38 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| RFC5 | |
| Polyclonal | |
| Unconjugated | |
| RFC5 | |
| 2610020K06Rik; 2610209F07Rik; 36.5kDa; 36kDa; A1 36 kDa subunit; activator 1 36 kDa subunit; Activator 1 subunit 5; MGC1155; Recc5; replication factor C (activator 1) 5; replication factor C (activator 1) 5, 36.5kDa; replication factor C 36 kDa subunit; replication factor C 5; replication factor C subunit 5; replication factor c subunit 5-like protein; replication factor C subunit RFC5; replication factor C, 36 kDa; replication factor C, 36-kDa subunit; RF-C 36 kDa subunit; RFC, 36.5 kD subunit; RFC36; Rfc5; rfc5 protein; Unknown (protein for MGC:128229); zgc:110313 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5985, 72151 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40937, Q9D0F6 | |
| RFC5 | |
| A synthesized peptide derived from human RFC5(Accession P40937), corresponding to amino acid residues L71-L121. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction