missing translation for 'onlineSavingsMsg'
Learn More

RFC5 Antibody (1B1), Novus Biologicals™

Product Code. 18398269 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398269 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398269 Supplier Novus Biologicals Supplier No. H00005985M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RFC5 Monoclonal antibody specifically detects RFC5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC5
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1B1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13961
Gene Alias Activator 1 36 kDa subunit, Activator 1 subunit 5,36.5 kD subunit, MGC1155, replication factor C (activator 1) 5, 36.5kDa, RF-C 36 kDa subunit, RFC36replication factor C (activator 1) 5 (36.5kD)
Host Species Mouse
Immunogen RFC5 (AAH13961, 1 a.a. ~ 340 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. METSALKQQEQPAATKIRNLPWVEKYRPQTLNDLISHQDILSTIQKFINEDRLPHLLLYGPPGTGKTSTILACAKQLYKDKEFGSMVLELNASDDRGIDIIRGPILSFASTRTIFKKGFKLVILDEADAMTQDAQNALRRVIEKFTENTRFCLICNYLSKIIPALQSRCTRFRFGPLTPELMAPRLEHVVEEEKVDISEDGMKALVTLSSGDMRRALNILQSTNMAFGKVTEETVYTCTGHPLKSDIANILDWMLNQDFTTAYRNITELKTLKGLALHDILTEIHLFVHRVDFPSSVRIHLLTKMADIEYRLSVGTNEKIQLSSLIAAFQVTRDLIVAEA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5985
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.