missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583954
This item is not returnable.
View return policy
Description
Immunogen sequence: RIIEPLTSRC SKFRFKPLSD KIQQQRLLDI AKKENVKISD EGIAYLVKVS EGDLRKAITF LQSATRLTGG KEITEKVITD IAGVIP Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 91%.
Human Replication Factor C (RFC) is a hetero-pentameric protein consisting of the p140, p40, p38, p37 and p36 subunits. RFC functions in the loading of the DNA polymerase delta processivity factor (PCNA, Proliferating Cell nuclear Antigen) onto a primed DNA template during DNA synthesis and repair. RFC may also participate in the DNA damage checkpoint response with a PCNA-like hetero-trimer consisting of Hus 1, Rad 1 and Rad 9.
Specifications
| RFC4 | |
| Polyclonal | |
| Unconjugated | |
| RFC4 | |
| A1; A1 37 kDa subunit; Activator 1 37 kDa subunit; activator 1 subunit 4; AI894123; AU040575; EGK_12194; replication factor C (activator 1) 4; replication factor C (activator 1) 4, 37kDa; replication factor C 37 kDa subunit; Replication factor C subunit 4; RFC 37 kDa subunit; RF-C 37 kDa subunit; RFC p37; RFC37; RFC4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35249 | |
| RFC4 | |
| Recombinant protein corresponding to Human RFC4. Recombinant protein control fragment (Product #RP-104105). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction