Learn More
RFC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-33083
Product Code. 18215860
  • £494.00 / 0.10mL
Description
RFC4 Polyclonal specifically detects RFC4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RFC4
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
RFC4
Recombinant protein encompassing a sequence within the center region of human RFC4. The exact sequence is proprietary.
0.1 mL
Cell Cycle and Replication, DNA Repair
5984
Human, Mouse
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.62mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
A1, A1 37 kDa subunit, Activator 1 37 kDa subunit, Activator 1 subunit 4, MGC27291, replication factor C (activator 1) 4, 37kDa, Replication factor C 37 kDa subunit, replication factor C subunit 4, RFC 37 kDa subunit, RF-C 37 kDa subunit, RFC37replication factor C (activator 1) 4 (37kD)
Rabbit
Affinity Purified
RUO
Primary
Tested in human. Mouse cross reactivity determined by western blot. Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.