missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11814-1-AP-20UL
This item is not returnable.
View return policy
Description
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 38 kD subunit. This subunit is essential for the interaction between the 140 kD subunit and the core complex that consists of the 36, 37, and 40 kD subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
| RFC3 | |
| Polyclonal | |
| Unconjugated | |
| RFC3 | |
| A1 38 kDa subunit, Activator 1 38 kDa subunit, Activator 1 subunit 3, Replication factor C subunit 3, RF C 38 kDa subunit, RFC3, RFC38 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288414, 5983, 69263 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P40938, Q8R323 | |
| Rfc3 | |
| RFC3 Fusion Protein Ag2269 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction