missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFC3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17702284
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17702284 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17702284 Supplier Invitrogen™ Supplier No. PA5120882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF-7, SW480, HL-60, NIH/3T3, 293T, Mouse liver. The target is usually found in the following locations: Nucleus. Immunogen sequence: MSLWVDKYRP CSLGRLDYHK EQAAQLRNLV QCGDFPHLLV YGPSGAGKKT RIMCILRELY GVGVEKLRIE HQTITTPSKK KIEISTIASN YHLEVNPSDA GNSDRVVIQE MLKTVAQSQQ LETNSQRDFK VVLLTEVDKL TKDAQHALRR TMEKYMSTCR LILCCNSTSK VIPPIRSRCL AVRVPAPSIE DICHVLSTVC KKEGLNLPSQ LAHRLAEKSC RNLRKALLMC EACRVQQYPF TADQEIPETD WEVYLRETAN AIVSQQTPQR LLEVRGRLYE.

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 38 kD subunit. This subunit is essential for the interaction between the 140 kD subunit and the core complex that consists of the 36, 37, and 40 kD subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RFC3
Gene Accession No. P40938, Q8R323
Gene Alias 2810416I22Rik; 38kDa; A1 38 kDa subunit; Activator 1 38 kDa subunit; activator 1 subunit 3; AU022547; DNA-binding; MGC5276; Polymorphism; Recc3; replication factor C (activator 1) 3; replication factor C (activator 1) 3, 38kDa; replication factor C 3; replication factor C 38 kDa subunit; Replication factor C subunit 3; replication factor C, 38 kDa; RF-C 38 kDa subunit; RFC, 38 kD subunit; RFC3; RFC38
Gene Symbols RFC3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288414, 5983, 69263
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.